Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neese Magnum 45 Rainwear: Bib Trousers with Safety Fly

Click to view available options
Size:
Medium
Large
X-Large
Description
- Neoprene on nylon
- Flame Resistant (ASTM D6413)
- Liquid Penetration (ASTM F903)
- Bib front coverage
- Adjustable elastic suspenders
- Non-conductive snap closures that fastens suspenders onto garment
- Safety fly with snap closure
- Suggested Applications: Industrial Wear, Sewer and Water Works, Pressure Wash Cleaning, Sanitation, Agriculture, Mining Wear
Specifications
Specifications
Color | Yellow |
Size | Medium |
Suspender | Elastic |
Size (English) Chest | 36 to 38 in. |
Size (English) Waist | 36 to 38 in. |
Size (Metric) Chest | 91.4 to 96.5 cm |
Style | Bib Trousers w/safety fly |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction