Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nefazodone hydrochloride, 98+%

Catalog No. aaj6279303
Click to view available options
Quantity:
1 g

CAS: 82752-99-6 | C25H33Cl2N5O2 | 506.47 g/mol

An antidepressant acts by modifying serotonin transmission. Mixed 5-HT2A serotonin receptor antagonist/serotonin uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An antidepressant acts by modifying serotonin transmission
  • Mixed 5-HT2A serotonin receptor antagonist/serotonin uptake inhibitor
TRUSTED_SUSTAINABILITY

Specifications

CAS 82752-99-6
Melting Point 186°C to 187°C
Color White
Molecular Formula C25H33Cl2N5O2
Quantity 1 g
MDL Number MFCD00935760
Merck Index 14,6438
Synonym nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
InChI Key DYCKFEBIOUQECE-UHFFFAOYSA-N
SMILES [H+].[Cl-].CCC1=NN(CCCN2CCN(CC2)C2=CC=CC(Cl)=C2)C(=O)N1CCOC1=CC=CC=C1
IUPAC Name hydrogen 1-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-3-ethyl-4-(2-phenoxyethyl)-4,5-dihydro-1H-1,2,4-triazol-5-one chloride
Molecular Weight (g/mol) 506.47
PubChem CID 54911
ChEBI CHEBI:7495
Formula Weight 506.47
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Nefazodone hydrochloride
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.