Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEFH (phospho KSP/KEP) Mouse anti-Human, Pig, Rat, Clone: NE14, Abnova™

Catalog No. 89034463 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-463 Supplier Abnova Corporation Supplier No. MAB14381
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native pig NEFH.

  • High specificity
  • High reactivity

Specifications

Antigen NEFH (phospho KSP/KEP)
Applications Flow Cytometry, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone NE14
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native pig NEFH.
Dilution Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Host Species Mouse
Immunogen Crude neurofilament preparation from porcine spinal cord.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This monoclonal antibody identified as heavy subunit of neurofilaments (NEFH). It reacts specifically with the phosphorylated KSP/KEP segment at the C-terminus of the heavy subunit of neurofilaments. After dephosphorylation of neurofilaments with alkaline phosphatase, this antibody no longer binds.
Target Species Human, Pig, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.