Learn More
Invitrogen™ NEFL Polyclonal Antibody

Description
For Western blot using alkaline phosphatase plus chromagen, a dilution of at least 1:1000 is recommended. For chemiluminescence, use at a dilution of 1:20,000. Stable at 4°C for at least one year. Mix 1:1 with 100% glycerol and store at -20°C for longer term storage.
Specifications
Specifications
| Antigen | NEFL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 5mM sodium azide |
| Gene | NEFL |
| Gene Accession No. | P02547, P02548, P07196, P08551, P19527 |
| Gene Alias | 68 kDa neurofilament protein; AI847934; CMT1F; CMT2E; FLJ53642; light molecular weight neurofilament protein; micro glutamic acid-rich protein; NEF3; NEFL; nefl.L; neurofilament; neurofilament 3; neurofilament L subunit; neurofilament light; neurofilament light polypeptide; neurofilament protein; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; neurofilament triplet L protein; neurofilament, light L homeolog; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament, light polypeptide L homeolog; neurofilament-l; neurofilament-light; Nf68; NFL; N-FL; NF-L; NFM; NF-M; NLC; OTTHUMP00000196614; PPP1R110; protein phosphatase 1, regulatory subunit 110; XELAEV_18018673mg; XNF-L |
| Gene Symbols | NEFL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.