Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEFM Mouse anti-Human, Clone:1B6B12, Abnova™

Catalog No. 89334520 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-520 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-520 Supplier Abnova Corporation Supplier No. MAB17835
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human NEFM.

Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and functionally maintain neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene encodes the medium neurofilament protein. This protein is commonly used as a biomarker of neuronal damage. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]

Specifications

Antigen NEFM
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1B6B12
Conjugate Unconjugated
Description neurofilament, medium polypeptide
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry (1:50-1:200)
Immunohistochemistry
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene NEFM
Gene Alias NEF3, NF-M, NFM
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 779-916 of human NEFM from E. coli.
Quantity 100 μg
Gene ID (Entrez) 4741
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.