Learn More
NEFM Mouse anti-Human, Clone: 2G12, Abnova™
Mouse monoclonal antibody raised against partial recombinant human NEFM.
Supplier: Abnova Corporation MAB12268
Description
Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and functionally maintain neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene encodes the medium neurofilament protein. This protein is commonly used as a biomarker of neuronal damage. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Specifications
NEFM | |
Monoclonal | |
Unconjugated | |
NEF3, NFM, NFM | |
Mouse | |
100 μg | |
Primary | |
Human | |
Purified |
ELISA, Flow Cytometry, Western Blot | |
2G12 | |
In PBS (0.05% sodium azide and 0.5% protein stabilizer) | |
NEFM | |
Recombinant protein corresponding to amino acids 381-443 of human NEFM. | |
RUO | |
4741 | |
Store at 4°C. For long term storage store at −20°C. Aliquot to avoid repeated freezing and thawing. | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.