Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Negative Control for Mouse Monoclonal Antibody, Clone: IGG1/453, Novus Biologicals™
SDP

Catalog No. p-7244778 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177761 20 μg
NB177760 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177761 Supplier Novus Biologicals Supplier No. NBP31390220UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

To serve as a true negative control for various procedures, this rabbit monoclonal IgG should be used under the identical conditions and at the same dilution as the rabbit monoclonal antibody being used for a positive reaction.

Specifications

Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone IGG1/453
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1-2 ug/ml, Immunohistochemistry-Frozen
Formulation 10 mM PBS with 0.05% BSA
Host Species Mouse
Immunogen Recombinant full-length human Negative Control for Mouse Monoclonal protein
Purification Method Protein A purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.