Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Negative Control for Mouse Monoclonal Antibody (IGG1/453), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314085UV
This item is not returnable.
View return policy
Description
To serve as a true negative control for various procedures, this rabbit monoclonal IgG should be used under the identical conditions and at the same dilution as the rabbit monoclonal antibody being used for a positive reaction.Specifications
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| IGG1/453 | |
| 50mM Sodium Borate | |
| Recombinant full-length human Negative Control for Mouse Monoclonal protein | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Monoclonal | |
| DyLight 350 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction