Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Negative Control for Rabbit Monoclonal Antibody (NCRBM/1520R), PE/Cy7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP254471PECY7
Description
Negative Control for Rabbit Monoclonal Monoclonal specifically detects Negative Control for Rabbit Monoclonal in All Species samples. It is validated for Flow Cytometry.Specifications
Negative Control for Rabbit Monoclonal | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
Recombinant full-length human KRT7 protein | |
0.1 mL | |
To serve as a true negative control for various procedures, this rabbit monoclonal IgG should be used under the identical conditions and at the same dilution as the rabbit monoclonal antibody being used for a positive reaction. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
NCRBM/1520R | |
Flow Cytometry | |
Rabbit | |
Protein A purified | |
Primary | |
Human, All species | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction