Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEGR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552870
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552870 Supplier Invitrogen™ Supplier No. PA552870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGGDKWSVDP RVSISTLNKR DYSLQIQNVD VTDDGPYTCS VQTQHTPRTM QVHLTVQVPP KIYDISNDMT VNEGTNVTLT CLATGKPEPS ISWRHISPSA KPFENGQYLD IYGITRDQAG EYECSA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

May be involved in cell-adhesion. May function as a trans-neural growth-promoting factor in regenerative axon sprouting in the mammalian brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEGR1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEGR1
Gene Accession No. Q7Z3B1
Gene Alias 5330422G01Rik; a kindred of IgLON; DMML2433; IgLON family member 4; IGLON4; Kiaa3001; KILON; kilon protein; kindred of IgLON; NEGR1; Neuronal growth regulator 1; neurotractin; Ntra; UNQ2433; UNQ2433/PRO4993
Gene Symbols NEGR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEGR1. Recombinant protein control fragment (Product #RP-89937).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257194
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.