Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEGR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585229 Supplier Invitrogen™ Supplier No. PA585229
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (95%), Rat (95%), Chicken (84%), Rhesus Monkey (98%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May be involved in cell-adhesion. May function as a trans-neural growth-promoting factor in regenerative axon sprouting in the mammalian brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEGR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene NEGR1
Gene Accession No. Q7Z3B1, Q80Z24
Gene Alias 5330422G01Rik; a kindred of IgLON; DMML2433; IgLON family member 4; IGLON4; Kiaa3001; KILON; kilon protein; kindred of IgLON; NEGR1; Neuronal growth regulator 1; neurotractin; Ntra; UNQ2433; UNQ2433/PRO4993
Gene Symbols NEGR1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NEGR1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257194, 320840
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.