Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEIL1 Antibody (OTI1D1), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272932APC
Description
NEIL1 Monoclonal antibody specifically detects NEIL1 in Human samples. It is validated for Western BlotSpecifications
NEIL1 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
OTI1D1 | |
Western Blot | |
DNA-(apurinic or apyrimidinic site) lyase Neil1, EC 3.2.2.-, EC 4.2.99.18, endonuclease 8-like 1, endonuclease VIII, Endonuclease VIII-like 1, FLJ22402, FPG1DNA glycosylase/AP lyase Neil1, hFPG1, NEH1, nei endonuclease VIII-like 1 (E. coli), Nei homolog 1, NEI1, Nei-like protein 1 | |
Human recombinant protein fragment corresponding to amino acids 186-390 of human NEIL1 (NP_078884) produced in E.coli. | |
0.1 mL | |
Base Excision Repair, DNA Repair | |
79661 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction