Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NEIL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578580
Description
Positive Control: human NEIL1-transfected 293T Predicted Reactivity: Mouse (87%), Rat (87%), Bovine (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NEIL1 belongs to a class of DNA glycosylases homologous to the bacterial Fpg/Nei family. These glycosylases initiate the first step in base excision repair by cleaving bases damaged by reactive oxygen species and introducing a DNA strand break via the associated lyase reaction.
Specifications
NEIL1 | |
Polyclonal | |
Unconjugated | |
NEIL1 | |
2810450N13Rik; DNA endonuclease eight-like glycosylase 1; DNA glycosylase/AP lyase Neil1; DNA-(apurinic or apyrimidinic site) lyase Neil1; endonuclease 8-like 1; endonuclease VIII; endonuclease VIII-like 1; FLJ22402; FPG1; hFPG1; NEH1; nei endonuclease VIII-like 1; nei endonuclease VIII-like 1 (E. coli); nei homolog 1; nei like DNA glycosylase 1; Nei1; NEIL1; nei-like DNA glycosylase 1; nei-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79661 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.78 mg/mL | |
0.1M tris glycine with 10% glycerol, 0.1M glycine and 0.01% thimerosal; pH 7 | |
Q96FI4 | |
NEIL1 | |
Recombinant protein encompassing a sequence within the center region of human NEIL1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction