Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEIL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585591 Supplier Invitrogen™ Supplier No. PA585591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (87%), Rat (87%), Bovine (91%). Positive Control: 293T, 293T nuclear extract, NEIL1-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NEIL1 belongs to a class of DNA glycosylases homologous to the bacterial Fpg/Nei family. These glycosylases initiate the first step in base excision repair by cleaving bases damaged by reactive oxygen species and introducing a DNA strand break via the associated lyase reaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEIL1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene NEIL1
Gene Accession No. Q96FI4
Gene Alias 2810450N13Rik; DNA endonuclease eight-like glycosylase 1; DNA glycosylase/AP lyase Neil1; DNA-(apurinic or apyrimidinic site) lyase Neil1; endonuclease 8-like 1; endonuclease VIII; endonuclease VIII-like 1; FLJ22402; FPG1; hFPG1; NEH1; nei endonuclease VIII-like 1; nei endonuclease VIII-like 1 (E. coli); nei homolog 1; nei like DNA glycosylase 1; Nei1; NEIL1; nei-like DNA glycosylase 1; nei-like protein 1
Gene Symbols NEIL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NEIL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79661
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.