Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEIL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103829
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103829 Supplier Invitrogen™ Supplier No. PA5103829
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NEIL2.

Involved in base excision repair of DNA damaged by oxidation or by mutagenic agents. Has DNA glycosylase activity towards 5-hydroxyuracil and other oxidized derivatives of cytosine with a preference for mismatched double-stranded DNA (DNA bubbles). Has low or no DNA glycosylase activity towards thymine glycol, 2-hydroxyadenine, hypoxanthine and 8-oxoguanine. Has AP (apurinic/apyrimidinic) lyase activity and introduces nicks in the DNA strand. Cleaves the DNA backbone by beta-delta elimination to generate a single-strand break at the site of the removed base with both 3'- and 5'-phosphates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEIL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NEIL2
Gene Accession No. Q6R2P8, Q969S2
Gene Alias DNA glycosylase/AP lyase Neil2; DNA-(apurinic or apyrimidinic site) lyase Neil2; Endonuclease 8-like 2; endonuclease VIII-like 2; Gm1212; NEH2; nei endonuclease VIII-like 2; nei endonuclease VIII-like 2 (E. coli); nei homolog 2; nei like 2; nei like 2 (E. coli); nei like DNA glycosylase 2; NEI2; Neil2; nei-like DNA glycosylase 2; nei-like protein 2
Gene Symbols NEIL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NEIL2(Accession Q969S2), corresponding to amino acid residues G128-Y178.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252969, 382913
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.