Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEIL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566796 Supplier Invitrogen™ Supplier No. PA566796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EALFDSGLHPA VKVCQLTDEQ IHHLMKMIRD FSILFYRCRK AGLALSKHYK VYKRPNCGQC HCRITVC Highest antigen sequence identity to the following orthologs - mouse 81%, rat 84%.

DNA glycosylase which prefers single-stranded DNA (ssDNA), or partially ssDNA structures such as bubble and fork structures, to double-stranded DNA (dsDNA). In vitro, displays strong glycosylase activity towards the hydantoin lesions spiroiminodihydantoin (Sp) and guanidinohydantoin (Gh) in both ssDNA and dsDNA; also recognizes FapyA, FapyG, 5-OHU, 5-OHC, 5-OHMH, Tg and 8-oxoA lesions in ssDNA. No activity on 8-oxoG detected. Also shows weak DNA-(apurinic or apyrimidinic site) lyase activity. In vivo, appears to be the primary enzyme involved in removing Sp and Gh from ssDNA in neonatal tissues. Seems to be an important facilitator of cell proliferation in certain populations, for example neural stem/progenitor cells and tumor cells, suggesting a role in replication-associated DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEIL3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Neil3
Gene Accession No. Q8TAT5
Gene Alias AI449477; BC034753; C85903; DNA glycosylase FPG2; DNA glycosylase hFPG2; DNA glycosylase/AP lyase Neil3; Endonuclease 8-like 3; endonuclease VIII-like 3; FGP2; FLJ10858; FPG2; hFPG2; hNEI3; nei endonuclease VIII-like 3; nei endonuclease VIII-like 3 (E. coli); nei like 3; nei like 3 (E. coli); nei like DNA glycosylase 3; NEI3; Neil3; nei-like DNA glycosylase 3; nei-like protein 3; RGD1310562; ZGRF3; zinc finger, GRF-type containing 3
Gene Symbols Neil3
Host Species Rabbit
Immunogen Recombinant Human NEIL3. Recombinant protein control fragment (Product #RP-107453).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55247
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.