Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neisseria gonorrhoeae Lipooligosaccharide (LOS) Antibody (3F11) - Galbeta1-4Glcnac, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274993
Description
Neisseria lipooligosaccharide (LOS) Monoclonal specifically detects Neisseria lipooligosaccharide (LOS) in Bacteria samples. It is validated for Western Blot, ELISA, Dot Blot.Specifications
Neisseria lipooligosaccharide (LOS) | |
Monoclonal | |
1 mg/ml | |
Western Blot, ELISA, Dot Blot | |
2-Acetamido-2-deoxy-4-O-beta-D-galactopyranosyl-D-glucopyranose, Gal-beta 1-4GlcNAc, LacNAc, N-Acetyl-4-O-(beta-D-galactopyranosyl)-D-glucosamine, N-Acetyl-D-Lactosamine | |
lactosamine (Galbeta1-4Glcnac) epitope found in lipooligosaccharide (LOS) of pathogenic Neisseria species | |
0.1 mL | |
Primary | |
Bacteria | |
IgM |
Western Blot, ELISA, Dot Blot | |
3F11 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Mouse | |
Thiophilic Chromatography | |
Immunology, Virology Bacteria and Parasites | |
Shown to recognize common antigenic region in gonococcal lipopolysaccharide (LPS). Recognizes conserved region on 6 different different gonococcal serotypes (90% of gonococci can be typed to 1 of these 6 serotypes) | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction