Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neisseria gonorrhoeae Lipooligosaccharide (LOS) Antibody (4C4) - Glcb1-4Hep - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274994
Description
Neisseria lipooligosaccharide (LOS) Monoclonal specifically detects Neisseria lipooligosaccharide (LOS) in Bacteria samples. It is validated for N/A.Specifications
Neisseria lipooligosaccharide (LOS) | |
4C4 | |
Unconjugated | |
beta -D-Glcp-(1->4)-L-alpha-D-Hepp, Glc-beta 1-4Hep, Glc-Hep | |
Glcb1-4Hep epitope found on N. meningitidis strain A1 and N. gonorrhoeae 1291c and 1291d strains | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Monoclonal | |
1 mg/ml | |
PBS with 0.02% Sodium Azide | |
Mouse | |
Protein A purified | |
Immunology, Virology Bacteria and Parasites | |
This antibody recognizes an epitope found on N. meningitidis strain A1 and N. gonorrhoeae 1291c and 1291d strains | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction