Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neisseria gonorrhoeae MOMP Antibody (386/418), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064715UV
Description
Neisseria gonorrhoeae MOMP Monoclonal antibody specifically detects Neisseria gonorrhoeae MOMP in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).Specifications
Neisseria gonorrhoeae MOMP | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Major outer membrane proteins (MOMP) from a mixture of detergent-treated Neisseria gonorrhoeae strains: SB587, SB2262, SB2265 & SB2275. | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
386/418 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
Mouse | |
Protein A purified | |
RUO | |
Reacts with a 30-31.5 kD MOMP protein of N. gonorrhoeae. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction