Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neisseria gonorrhoeae MOMP Monoclonal Antibody (386/418)
GREENER_CHOICE

Catalog No. PIMA183358
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183358 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183358 Supplier Invitrogen™ Supplier No. MA183358
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The immunogen used to produce MA1-83358 is major outer membrane proteins (MOMP) from a mixture of detergent-treated Neisseria gonorrhoeae strains: SB587, SB2262, SB2265, & SB2275. Mouse anti Neisseria gonorrhoeae MOMP 30 kDa antibody, clone 386/418 reacts with the 30-31.5 kDa major outer membrane protein (MOMP) of Neisseria gonorrhoeae a gram-negative diplococcus known to be the causative agent of sexually transmitted gonorrhoea.

The genus Neisseria contains a number of species which are normal flora and pathogens of humans and animals. Of these species, the species of human origin, and particularly the pathogenic species, N. gonorrhoeae and N. meningitidis, have been studied extensively in an effort to control the infections they cause. Gonorrhea, caused by N. gonorrhoeae, is one of the most frequently reported infectious diseases in the United States and worldwide. Of the Neisseria and related species, only N. gonorrhoeae is considered always to be pathogenic i.e., cause disease. N. gonorrhoeae is not considered to be normal flora under any circumstances.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neisseria gonorrhoeae MOMP
Applications ELISA
Classification Monoclonal
Clone 386/418
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias Gonococci; N. gonorrhoeae
Host Species Mouse
Immunogen Major outer membrane proteins (MOMP) from a mixture of detergent-treated Neisseria gonorrhoeae strains: SB587, SB2262, SB2265, & SB2275.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.