Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neisseria gonorrhoeae Polyclonal Antibody, HRP
GREENER_CHOICE

Catalog No. PA173144
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173144 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173144 Supplier Invitrogen™ Supplier No. PA173144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73144 antibody reacts with Neisseria gonorrhoeae. PA1-73144 has been successfully used in ELISA, IHC, and IB applications. The PA1-73144 antibody was raised against whole N. gonorrhoeae; ATCC 31426.

Although pathogenic bacteria constitute only a small portion of bacterial species, infectious diseases caused by pathogenic bacteria affect millions of people worldwide. Diarrheal diseases, many of which are bacterial, are the second leading cause of death worldwide, claiming an estimated 5 million people annually. Furthermore, an estimated 3 million people die of tuberculosis annually, 500,000 people die of pertussis, and 25,000 die of typhoid. The ability of bacteria to utilize various food sources is a major factor behind their pervasiveness. Their vast numbers lead to rapid evolutionary changes resulting in emerging diseases as well as antibiotic-resistant strains of well known diseases. Examples of these are multi-drug resistant tuberculosis and methicillin-resistant Staphylococcus aureus. Research into the detection and treatment of these infectious disease agents will continue to expand in an effort to alleviate the effects of these diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neisseria gonorrhoeae
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1-2 mg/mL
Conjugate HRP
Formulation PBS with 10mg/mL BSA and 0.002% thimerosal; pH 7.2
Gene Alias Gonococci; N. gonorrhoeae
Host Species Rabbit
Immunogen Whole N. gonorrhoeae; ATCC 31426.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.