Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neisseria meningitidis Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA173149
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173149 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173149 Supplier Invitrogen™ Supplier No. PA173149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73149 antibody reacts with Neisseria meningitidis. PA1-73149 has been successfully used in Immunofluorescence applications. The PA1-73149 antibody was raised against whole cell prep. of N. meningitidis types A, B & C.

Although pathogenic bacteria constitute only a small portion of bacterial species, infectious diseases caused by pathogenic bacteria affect millions of people worldwide. Diarrheal diseases, many of which are bacterial, are the second leading cause of death worldwide, claiming an estimated 5 million people annually. Furthermore, an estimated 3 million people die of tuberculosis annually, 500,000 people die of pertussis, and 25,000 die of typhoid. The ability of bacteria to utilize various food sources is a major factor behind their pervasiveness. Their vast numbers lead to rapid evolutionary changes resulting in emerging diseases as well as antibiotic-resistant strains of well known diseases. Examples of these are multi-drug resistant tuberculosis and methicillin-resistant Staphylococcus aureus. Research into the detection and treatment of these infectious disease agents will continue to expand in an effort to alleviate the effects of these diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neisseria meningitidis
Applications Immunocytochemistry
Classification Polyclonal
Concentration 4-5 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Meningococcus
Host Species Rabbit
Immunogen Whole cell prep. of N. meningitidis types A, B & C.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.