Learn More
Invitrogen™ Neisseria meningitidis Polyclonal Antibody, FITC

Description
The PA1-73147 antibody reacts with Neisseria meningitidis. PA1-73147 has been successfully used in Immunofluorescence applications. The PA1-73147 antibody was raised against whole cell prep. of N. meningitidis types A, B & C.
Specifications
Specifications
| Antigen | Neisseria meningitidis |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 |
| Gene Alias | Meningococcus |
| Host Species | Rabbit |
| Immunogen | Whole cell prep. of N. meningitidis types A, B & C. |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.