Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559166 Supplier Invitrogen™ Supplier No. PA559166
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLKEQLERKR KEAYEREKKV WEEHLVAKGV KSSDVSPPLG QHETGGSPSK QQMRSVISVT SALKEVGVDS SLTDTRETSE EMQKTNNAIS SKR Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.

NEK1 is a serine/threonine protein kinase, related to the NIMA cell cycle regulator, and is highly expressed in meiotic germ cells. The kinase may function early in the DNA damage response pathway. NEK1 may also participate in polycystic kidney disease and neural cell development. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEK1
Gene Accession No. Q96PY6
Gene Alias D8Ertd790e; DKFZp686D06121; DKFZp686K12169; kat; KIAA1901; kidney, anemia and testis; MGC138800; NEK1; Nek1 serine/threonine- and tyrosine-specific protein kinase; never in mitosis A-related kinase 1; NIMA (never in mitosis gene a)-related expressed kinase 1; NIMA (never in mitosis gene a)-related kinase 1; NIMA related kinase 1; NIMA-related kinase 1; nimA-related protein kinase 1; NY\ REN55; NY-REN-55; NY-REN-55 antigen; protein-serine/threonine kinase; renal carcinoma antigen NY-REN-55; Serine/threonine-protein kinase Nek1; SRPS2; SRPS2A; SRTD6
Gene Symbols NEK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEK1. Recombinant protein control fragment (Product #RP-98309).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4750
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.