Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK10 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299009F
Description
NEK10 Polyclonal antibody specifically detects NEK10 in Human samples. It is validated for Western BlotSpecifications
NEK10 | |
Polyclonal | |
Western Blot | |
FLJ32685, NIMA (never in mitosis gene a)- related kinase 10 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NEK10. | |
0.1 mL | |
Protein Kinase | |
152110 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction