Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEK10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558601 Supplier Invitrogen™ Supplier No. PA558601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHILQGDRNF VSDHSSIGSL SSANAAGRIQ QLHLSEDLSP REIQENTFSL QAACCAALTE LVLNDTNAHQ VVQENGVYTI AKLILPNKQK NAAKSNLLQC YAFRALRFLF SMERNRPLFK RLFPTDLF Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

NEK10 (NIMA Related Kinase 10) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includetransferase activity, transferring phosphorus-containing groupsandprotein tyrosine kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEK10
Gene Accession No. Q6ZWH5
Gene Alias FLJ32685; NEK10; Never in mitosis A-related kinase 10; NIMA (never in mitosis gene a)- related kinase 10; NIMA related kinase 10; NIMA-related kinase 10; nimA-related protein kinase 10; Serine/threonine-protein kinase Nek10
Gene Symbols NEK10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEK10. Recombinant protein control fragment (Product #RP-89824).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152110
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.