Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NEK10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581225
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human NEK10.
NEK10 (NIMA Related Kinase 10) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includetransferase activity, transferring phosphorus-containing groupsandprotein tyrosine kinase activity.
Specifications
NEK10 | |
Polyclonal | |
Unconjugated | |
NEK10 | |
FLJ32685; NEK10; Never in mitosis A-related kinase 10; NIMA (never in mitosis gene a)- related kinase 10; NIMA related kinase 10; NIMA-related kinase 10; nimA-related protein kinase 10; Serine/threonine-protein kinase Nek10 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
152110 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q6ZWH5 | |
NEK10 | |
A synthetic peptide corresponding to the N-terminus of the Human NEK10. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction