Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK10 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549478
Description

Specifications
NEK10 | |
Polyclonal | |
Unconjugated | |
NEK10 | |
Serine/threonine-protein kinase Nek10, Never in mitosis A-related kinase 10, NimA-related protein kinase 10, NEK10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
152110 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide | |
Q6ZWH5 | |
NEK10 | |
KLH conjugated synthetic peptide between 450-483 amino acids from the Central region of human NEK10. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction