Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK11 Antibody (OTI4H5), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272933AF405
Description
NEK11 Monoclonal specifically detects NEK11 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NEK11 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NEK11 | |
Full length recombinant protein of human NEK11 (NP_079076) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI4H5 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
EC 2.7.11, EC 2.7.11.1, FLJ23495, Never in mitosis A-related kinase 11, NIMA (never in mitosis gene a)- related kinase 11, NimA-related protein kinase 11, serine/threonine-protein kinase Nek11 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
79858.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction