Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK11 Antibody (OTI5D9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246140
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
NEK11 Monoclonal specifically detects NEK11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
NEK11 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
EC 2.7.11, EC 2.7.11.1, FLJ23495, Never in mitosis A-related kinase 11, NIMA (never in mitosis gene a)- related kinase 11, NimA-related protein kinase 11, serine/threonine-protein kinase Nek11 | |
Mouse | |
74 kDa | |
0.1 mL | |
Protein Kinase | |
79858 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunofluorescence | |
OTI5D9 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
NEK11 | |
Full length human recombinant protein of human NEK11(NP_079076) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction