Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK11 Monoclonal Antibody (OTI5D9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501765S


Description
NEK11 Monoclonal antibody specifically detects NEK11 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
NEK11 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q6ZUJ8 | |
Mouse | |
Affinity chromatography | |
RUO | |
118788, 294048 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI5D9 | |
Unconjugated | |
PIK3AP1 | |
4932416N14Rik; Nek11; never in mitosis A-related kinase 11; NIMA (never in mitosis gene a)- related kinase 11; NIMA (never in mitosis gene a)-related expressed kinase 11; NIMA related kinase 11; NIMA-related kinase 11; nimA-related protein kinase 11; Serine/threonine-protein kinase Nek11 | |
Full length human recombinant protein of human PIK3AP1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction