Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553516 Supplier Thermo Scientific Supplier No. PA553516
Add to Cart
Add to Cart

NEK11 Polyclonal Antibody

NEK11 Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK11
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nek11
Gene Accession No. Q8NG66
Gene Alias 4932416N14Rik; Nek11; never in mitosis A-related kinase 11; NIMA (never in mitosis gene a)- related kinase 11; NIMA (never in mitosis gene a)-related expressed kinase 11; NIMA related kinase 11; NIMA-related kinase 11; nimA-related protein kinase 11; Serine/threonine-protein kinase Nek11
Gene Symbols Nek11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEK11. Recombinant protein control fragment (Product #RP-89776).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79858
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.