Learn More
Promega NEK2 Kinase Enzyme System, 1mg
Catalog No. PRV3872 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3872 1 mg
1 options
Catalog No. PRV3872 Supplier Promega Supplier No. V3872
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human NEK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NEK2 is closely related in its catalytic domain to the serine/threonine protein kinase NIMA of Aspergillus nidulans.
  • Easily Screen and Profile NEK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg