Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NEK2 Monoclonal Antibody (1B6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA134594
Description
Recommended positive controls: Jurkat cell lysate.
NEK2 is involved in cell cycle regulation and is essential for faithful chromosome segregation. NEK2 is a gene that encodes for a serine/threonine-protein kinase. This protein plays a crucial role in the regulation of mitosis and is primarily located in the centrosome. During the G1 phase, the protein is undetectable, but its levels progressively increase throughout the S phase, reaching their peak in late G2 phase. The gene has several alternatively spliced transcript variants that encode different isoforms with distinct C-termini. NEK2 is associated with several diseases, including Retinitis Pigmentosa 67 and Retinitis Pigmentosa.
Specifications
NEK2 | |
Monoclonal | |
0.43 mg/mL | |
PBS with no preservative | |
P51955 | |
NEK2 | |
Synthetic peptide ETH; Immunogen sequence information is commercially sensitive. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
1B6 | |
Unconjugated | |
NEK2 | |
AA617254; C77054; HSPK 21; HsPK21; NEK 2; NEK 2A; Nek2; Nek2 kinase; NEK2A; Never in mitosis A-related kinase 2; NIMA - related expressed kinase 2; NIMA (never in mitosis gene a)-related expressed kinase 2; NIMA (never in mitosis gene a)-related kinase 2; NIMA related kinase 2; nimA-like protein kinase 1; NIMA-related expressed kinase 2; NIMA-related kinase 2; nimA-related protein kinase 2; NLK 1; NLK1; PPP1R111; protein phosphatase 1, regulatory subunit 111; RP67; Serine/threonine-protein kinase Nek2 | |
Mouse | |
Protein G | |
RUO | |
4751 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction