Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK2 Mouse anti-Human, Mouse, Rat, Clone: 3D7B4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | NEK2 |
---|---|
Clone | 3D7B4 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7050
![]() ![]() |
Proteintech Group Inc
666321IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a serine/threonine-protein kinase that is involved in mitotic regulation. This protein is localized to the centrosome, and undetectable during G1 phase, but accumulates progressively throughout the S phase, reaching maximal levels in late G2 phase.Specifications
NEK2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HSPK 21, HsPK21, NEK2, NEK2A, NimA like protein kinase 1, NimA related protein kinase 2, NLK1 | |
Nek2 | |
IgG1 | |
Protein G | |
Antibody |
3D7B4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
O35942, P51955 | |
114482, 18005, 4751 | |
NEK2 Fusion Protein Ag21476 | |
Primary | |
-20°C | |
NEK2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title