Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK2 Rabbit anti-Human, Mouse, Rat, Clone: 5O0A1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | NEK2 |
---|---|
Clone | 5O0A1 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
NEK2 Monoclonal antibody specifically detects NEK2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
NEK2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
4751 | |
IgG | |
Affinity purified |
5O0A1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.11, EC 2.7.11.1, HSPK 21, NEK2AHsPK21, Never in mitosis A-related kinase 2, NIMA (never in mitosis gene a)-related kinase 2, NimA-like protein kinase 1, NimA-related protein kinase 2, NLK1serine/threonine-protein kinase Nek2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NEK2 (P51955). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title