Learn More
Promega NEK3 Kinase Enzyme System, 1mg
Catalog No. PRV6592 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6592 1 mg
1 options
Catalog No. PRV6592 Supplier Promega Supplier No. V6592
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human NEK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NEK3 is a member of the NEK family of protein kinases that share high amino acid homology with NIMA (never in mitosis gene a).
  • Easily Screen and Profile NEK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C