Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528569
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528569 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528569 Supplier Invitrogen™ Supplier No. PA528569
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Mouse (89%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NIMA was originally shown in Aspergillus nidulans to be necessary for entry into mitosis. NIMA-related mammalian proteins have since been identified as Nek1, Nek2, Nek3 and Nek4 (also designated STK2 or NRK2). High expression of Nek1 is seen in male and female germ cell lines of mouse. Nek2 is the closest known mammalian relative to NIMA. Like NIMA, Nek2 expression peaks at the G2 to M phase transition. Nek3 is a predominantly cytoplasmic enzyme that was detectable in all organs studied. Levels of Nek3 seem to remain unchanged throughout the cell cycle, but appear to be elevated in G0-arrested, quiescent fibroblasts. In developing testicular germ cells, differential patterns of expression were seen for Nek1, Nek2 and Nek4, indicating possible overlapping, but non-identical functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Nek3
Gene Accession No. P51956
Gene Alias glycogen synthase A kinase; HSPK 36; HSPK36; hydroxyalkyl-protein kinase; Nek3; Never in mitosis A-related kinase 3; NIMA (never in mitosis gene a)-related expressed kinase 3; NIMA (never in mitosis gene a)-related kinase 3; NIMA related kinase 3; NIMA-related expressed kinase 3; NIMA-related kinase 3; nimA-related protein kinase 3; phosphorylase B kinase kinase; serine/threonine-protein kinase Nek3
Gene Symbols Nek3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NEK3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4752
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.