Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK4 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142686AF350
Description
NEK4 Polyclonal antibody specifically detects NEK4 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
NEK4 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
EC 2.7.11, Never in mitosis A-related kinase 4, NIMA (never in mitosis gene a)-related kinase 4, NimA-related protein kinase 4, NRK2EC 2.7.11.1, pp12301, serine/threonine kinase 2, serine/threonine protein kinase-2, Serine/threonine-protein kinase 2, serine/threonine-protein kinase Nek4, Serine/threonine-protein kinase NRK2, STK2MGC33171 | |
The immunogen for this product maps to a region between residue 350 and 400 of human NIMA (never in mitosis gene a)-related kinase 4 using the numbering given in entry NP_003148.2 (GeneID 6787). | |
0.1 mL | |
Cell Cycle and Replication | |
6787 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction