Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110022 Supplier Thermo Scientific Supplier No. PA5110022
Add to Cart
Add to Cart

NEK4 Polyclonal Antibody

NEK4 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NEK4
Gene Accession No. Q9Z1J2
Gene Alias Nek4; Never in mitosis A-related kinase 4; NIMA (never in mitosis gene a)-related expressed kinase 4; NIMA (never in mitosis gene a)-related kinase 4; NIMA related kinase 4; NIMA-related kinase 4; nimA-related protein kinase 4; NRK2; pp12301; serine/threonine kinase 2; serine/threonine protein kinase-2; serine/threonine-protein kinase 2; Serine/threonine-protein kinase Nek4; serine/threonine-protein kinase Nek4 variant 2; serine/threonine-protein kinase NRK2; STK2
Gene Symbols NEK4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human NEK4 (NP_003148.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23955, 306252
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.