Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101860 Supplier Invitrogen™ Supplier No. PA5101860
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NEK5.

NEK5 gene ontology (GO) annotations include positive regulation of cysteine-type endopeptidase activity; positive regulation of striated muscle cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NEK5
Gene Accession No. Q6P3R8, Q7TSC3
Gene Alias 6530411J22; BB049667; Nek5; Never in mitosis A-related kinase 5; NIMA (never in mitosis gene a)-related expressed kinase 5; NIMA (never in mitosis gene a)-related kinase 5; NIMA related kinase 5; NIMA-related kinase 5; nimA-related protein kinase 5; Serine/threonine-protein kinase Nek5
Gene Symbols NEK5
Host Species Rabbit
Immunogen A synthesized peptide derived from human NEK5(Accession Q6P3R8), corresponding to amino acid residues E159-L209.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 330721, 341676
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.