Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NEK5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559551
Description
Immunogen sequence: FQELPFRKNE MKEQEYWKQL EEIRQQYHND MKEIRKKMGR EPEENSKISH KTYLVKKSNL PVHQDASEGE APVQ Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 49%.
NEK5 gene ontology (GO) annotations include positive regulation of cysteine-type endopeptidase activity; positive regulation of striated muscle cell differentiation.
Specifications
NEK5 | |
Polyclonal | |
Unconjugated | |
NEK5 | |
6530411J22; BB049667; Nek5; Never in mitosis A-related kinase 5; NIMA (never in mitosis gene a)-related expressed kinase 5; NIMA (never in mitosis gene a)-related kinase 5; NIMA related kinase 5; NIMA-related kinase 5; nimA-related protein kinase 5; Serine/threonine-protein kinase Nek5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
341676 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P3R8 | |
NEK5 | |
Recombinant protein corresponding to Human NEK5. Recombinant protein control fragment (Product #RP-104268). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction