Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEK6 Monoclonal Antibody (OTI5D7)
GREENER_CHOICE

Catalog No. PIMA524952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524952 Supplier Invitrogen™ Supplier No. MA524952
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEK6 Monoclonal Antibody for Flow, ICC/IF, IHC (P)

NEK6 kinase is a member of the NEK family. NEK6 is a serine/threonine kinase that controls the initiation of mitosis and is required for the metaphase-anaphase transition. The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone OTI5D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NEK6
Gene Accession No. Q9HC98
Gene Alias 1300007C09Rik; Nek6; Never in mitosis A-related kinase 6; NIMA (never in mitosis gene a)-related expressed kinase 6; NIMA (never in mitosis gene a)-related kinase 6; NIMA related kinase 6; NIMA-related expressed kinase 6; NIMA-related kinase 6; nimA-related protein kinase 6; Protein kinase SID6-1512; putative serine-threonine protein kinase; RP11-101K10.6; serine/threonine-protein kinase Nek6; SID; SID6-1512
Gene Symbols NEK6
Host Species Mouse
Immunogen Full length human recombinant protein of NEK6 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10783
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.