Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEK6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566044 Supplier Invitrogen™ Supplier No. PA566044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVHPPDPQRHP NTLSFRCSLA DFQIE Highest antigen sequence identity to the following orthologs - mouse 88%, rat 96%.

NEK6 kinase is a member of the NEK family. NEK6 is a serine/threonine kinase that controls the initiation of mitosis and is required for the metaphase-anaphase transition. The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEK6
Gene Accession No. Q9HC98
Gene Alias 1300007C09Rik; Nek6; Never in mitosis A-related kinase 6; NIMA (never in mitosis gene a)-related expressed kinase 6; NIMA (never in mitosis gene a)-related kinase 6; NIMA related kinase 6; NIMA-related expressed kinase 6; NIMA-related kinase 6; nimA-related protein kinase 6; Protein kinase SID6-1512; putative serine-threonine protein kinase; RP11-101K10.6; serine/threonine-protein kinase Nek6; SID; SID6-1512
Gene Symbols NEK6
Host Species Rabbit
Immunogen Recombinant Human NEK6. Recombinant protein control fragment (Product #RP-105056).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10783
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.