Learn More
Invitrogen™ NEK6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597077
Description
Immunogen sequence: MAGQPGHMPH GGSSNNLCHT LGPVHPPDPQ RHPNTLSFRC SLADFQIEKK IGRGQFSEVY KATCLLDRKT VALKKVQIFE MMDAKARQDC VKEIGLLKQL NHPNIIKYLD SFIEDNELNI VLELADAGDL SQMIKYFKKQ KRLIPERTVW KYFVQLCSAV EHMHSRRVMH RDIKPANVFI TATGVVKLGD LGLGRFFSSE TTAAHSLVGT PYYMSPERIH ENGYNFKSDI WSLGCLLYEM AALQSPFYGD KMNLFSLCQK IEQCDYPPLP GEHYSEKLRE LVSMCICPDP HQRPDIGYVH QVAKQMHIWM SST; Positive Samples: 293T; Cellular Location: Cytoplasm, Nucleus, Nucleus speckle, centrosome, cytoskeleton, microtubule organizing center, spindle pole.
NEK6 kinase is a member of the NEK family. NEK6 is a serine/threonine kinase that controls the initiation of mitosis and is required for the metaphase-anaphase transition. The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.
Specifications
NEK6 | |
Polyclonal | |
Unconjugated | |
NEK6 | |
1300007C09Rik; Nek6; Never in mitosis A-related kinase 6; NIMA (never in mitosis gene a)-related expressed kinase 6; NIMA (never in mitosis gene a)-related kinase 6; NIMA related kinase 6; NIMA-related expressed kinase 6; NIMA-related kinase 6; nimA-related protein kinase 6; Protein kinase SID6-1512; putative serine-threonine protein kinase; RP11-101K10.6; serine/threonine-protein kinase Nek6; SID; SID6-1512 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10783 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.64 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HC98 | |
NEK6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 35-347 of human NEK6 (NP_0011596431). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.