Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEK7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101861 Supplier Invitrogen™ Supplier No. PA5101861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NEK7.

NEK7 (NIMA Related Kinase 7) is a Protein Coding gene. Diseases associated with NEK7 include Muckle-Wells Syndrome and Cinca Syndrome. Among its related pathways are Cell Cycle, Mitotic and Transport of the SLBP independent Mature mRNA. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is NEK6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NEK7
Gene Accession No. D3ZBE5, Q8TDX7, Q9ES74
Gene Alias 2810460C19Rik; AU020186; NEK7; Never in mitosis A-related kinase 7; NIMA (never in mitosis gene a)-related expressed kinase 7; NIMA (never in mitosis gene a)-related kinase 7; NIMA related kinase 7; NIMA-related expressed kinase 7; NIMA-related kinase 7; nimA-related protein kinase 7; Serine/threonine-protein kinase Nek7
Gene Symbols NEK7
Host Species Rabbit
Immunogen A synthesized peptide derived from human NEK7(Accession Q8TDX7), corresponding to amino acid residues S100-E150.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140609, 360850, 59125
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.