Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK7 Rabbit anti-Human, Mouse, Rat, Clone: 7Y3B10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | NEK7 |
---|---|
Clone | 7Y3B10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
NEK7 Monoclonal antibody specifically detects NEK7 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
NEK7 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Protein Kinase | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
140609 | |
IgG | |
Affinity purified |
7Y3B10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.11.1, Never in mitosis A-related kinase 7, NIMA (never in mitosis gene a)-related kinase 7, NimA-related protein kinase 7, serine/threonine-protein kinase Nek7 | |
A synthetic peptide corresponding to a sequence within amino acids 203-302 of human NEK7 (Q8TDX7). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title