Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEK8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559248 Supplier Invitrogen™ Supplier No. PA559248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSTERELFAW GRGDSGRLGL GTRESHSCPQ QVPMPPGQEA QRVVCGIDSS MILTVPGQAL ACGSNRFNKL G Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.

NEK8 serine/threonine kinase or NIMA-related kinase 8 is related to NIMA (never in mitosis, gene A) of Aspergillus nodulins and may play a role in cell cycle progression from G2 to M phase. NEK8 is part of a complex of ciliary proteins required for renal and cardiovascular development. NEK8 interacts with polycystin signal transduction pathways and may control the targeting of these ciliary proteins. Required for renal tubular integrity. May regulate local cytoskeletal structure in kidney tubule epithelial cells. May regulate ciliary biogenesis through targeting of proteins to the cilia.Defects in NEK8 are the cause of nephronophthisis type 9 (NPHP9) [MIM:613824]. NPHP9 is an autosomal recessive disorder resulting in end-stage renal disease. It is a progressive tubulo-interstitial kidney disorder histologically characterized by modifications of the tubules with thickening of the basement membrane, interstitial fibrosis and, in the advanced stages, medullary cysts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEK8
Gene Accession No. Q86SG6
Gene Alias 4632401F23Rik; b2b1449Clo; JCK; juvenile cystic kidney; NEK 8; NEK12A; Nek8; nephrocystin 9; never in mitosis A-related kinase 8; NIMA (never in mitosis gene a)- related kinase 8; NIMA (never in mitosis gene a)-related expressed kinase 8; NIMA related kinase 8; NIMA-family kinase NEK8; NIMA-related expressed kinase 8; NIMA-related kinase 12a; NIMA-related kinase 8; nima-related protein kinase 12a; NimA-related protein kinase 8; NPHP9; RHPD2; Serine/threonine-protein kinase Nek8; serine/thrionine-protein kinase NEK8
Gene Symbols NEK8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEK8. Recombinant protein control fragment (Product #RP-98305).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284086
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.