Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK9 Antibody (OTI1H1), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272939F
Description
NEK9 Monoclonal specifically detects NEK9 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
NEK9 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NEK9 | |
Full length recombinant protein of human NEK9 (NP_149107) produced in HEK293 cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1H1 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
DKFZp434D0935, KIAA1995, MGC138306, MGC16714, NEK8, Nek8NERCC1, Nercc1 kinase, NERCCEC 2.7.11.1, Never in mitosis A-related kinase 9, NIMA (never in mitosis gene a)- related kinase 9, NimA-related kinase 8, NIMA-related kinase Nek8, NimA-related protein kinase 9, serine/threonine-protein kinase Nek9 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, DNA Repair, GPCR, Phospho Specific | |
91754.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction