Learn More
Invitrogen™ NEK9 Monoclonal Antibody (OTI1H1)

Description
NEK9 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | NEK9 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1H1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NEK9 |
| Gene Accession No. | Q8TD19 |
| Gene Alias | APUG; C130021H08Rik; DKFZp434D0935; KIAA1995; LCCS10; MGC138306; MGC16714; NC; NEK 9; NEK8; NEK9; Nercc; Nercc 1 kinase; NERCC1; Nercc1 kinase; never in mitosis A-related kinase 9; NIMA (never in mitosis gene a)- related kinase 9; NIMA (never in mitosis gene a)-related expressed kinase 9; NIMA related kinase 9; NimA-related kinase 8; NIMA-related kinase 9; nimA-related protein kinase 9; Serine/threonine-protein kinase Nek9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.